Learn More
Medchemexpress LLC HY-100422 5mg Medchemexpress, KDM5-IN-1 CAS:1628210-26-3 Purity:>98%
SDP
Catalog No. 501931826
Encompass_Preferred
Medchemexpress, HY-100422 5mg KDM5-IN-1 CAS:1628210-26-3 KDM5-IN-1 is a potent, selective and orally bioavailable KDM5 inhibitor with an IC 50 of 15.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1826 Supplier Medchemexpress LLC Supplier No. HY1004225MG
Medchemexpress, HY-100422 5mg KDM5-IN-1 CAS:1628210-26-3 KDM5-IN-1 is a potent, selective and orally bioavailable KDM5 inhibitor with an IC 50 of 15.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.