Learn More
Sino Biological Recombinant Human KDM2A Protein (GST Tag)
Supplier: Sino Biological K42231G100
Recombinant human KDM2A (1-748) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.