Learn More
ABclonal Technology KDM5C Rabbit pAb
SDP
Supplier:  ABclonal Technology A157401000UL
Encompass_Preferred
This gene is a member of the SMCY homolog family and encodes a protein with one ARID domain, one JmjC domain, one JmjN domain and two PHD-type zinc fingers. The DNA-binding motifs suggest this protein is involved in the regulation of transcription and chromatin remodeling. Mutations in this gene have been associated with X-linked cognitive disability. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-05492