Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KDOAM-25 citrate | 2448475-08-7 | MFCD00865590 | >98.0% | 499.51 g/mol | C21H33N5O9 | 1 ML
SDP

Catalog No. 5000247374
Encompass_Preferred

KDOAM-25 citrate is the citrate salt of a potent, selective inhibitor of KDM5 family histone lysine demethylases. It is supplied as a solid or as a 10 mM solution in DMSO (1 mL) and is intended for use in biochemical and cellular research probing KDM5 activity and epigenetic regulation.

  • Potent, selective KDM5 inhibitor with low-nanomolar IC50 values.
  • Available as solid or 10 mM solution in DMSO (1 mL).
  • High purity (>98% by HPLC/LCMS).
  • Molecular weight 499.51 g/mol; formula C21H33N5O9.
  • Suitable for biochemical assays and cellular studies.
  • Soluble in DMSO for convenient stock preparation.

Catalog No. 50-002-47374 Supplier Medchemexpress LLC Supplier No. HY102047B10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KDOAM-25 citrate is the citrate salt of a potent, selective inhibitor of KDM5 family histone lysine demethylases. It is supplied as a solid or as a 10 mM solution in DMSO (1 mL) and is intended for use in biochemical and cellular research probing KDM5 activity and epigenetic regulation.

  • Potent, selective KDM5 inhibitor with low-nanomolar IC50 values.
  • Available as solid or 10 mM solution in DMSO (1 mL).
  • High purity (>98% by HPLC/LCMS).
  • Molecular weight 499.51 g/mol; formula C21H33N5O9.
  • Suitable for biochemical assays and cellular studies.
  • Soluble in DMSO for convenient stock preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.