Learn More
Medchemexpress LLC HY-102047B 5mg Medchemexpress, KDOAM-25 (citrate) CAS: Purity:>98%
SDP
Catalog No. 501967970
Encompass_Preferred
Medchemexpress, HY-102047B 5mg KDOAM-25 (citrate) CAS: KDOAM-25 citrate is a potent and highly selective KDM5 sub-family of histone lysine demethylases inhibitor with IC50s of 71 nM, 19 nM, 69 nM, 69 nM for KDM5A, KDM5B, KDM5C, KDM5D, respectively. KDOAM-25 citrate increases global H3K4 methylation at transcriptional start sites and impairs proliferation in multiple myeloma MM1S cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7970 Supplier Medchemexpress LLC Supplier No. HY102047B5MG
Medchemexpress, HY-102047B 5mg KDOAM-25 (citrate) CAS: KDOAM-25 citrate is a potent and highly selective KDM5 sub-family of histone lysine demethylases inhibitor with IC50s of 71 nM, 19 nM, 69 nM, 69 nM for KDM5A, KDM5B, KDM5C, KDM5D, respectively. KDOAM-25 citrate increases global H3K4 methylation at transcriptional start sites and impairs proliferation in multiple myeloma MM1S cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.