Learn More
Carnabio Usa Inc KDR/5UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 081915
Encompass
KDR, Cytoplasmic domain, Wild type, Amino Acid 790-1356, NP_002244.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0477