Learn More
Aviva Systems Biology Corp KDR Antibody - N-terminal region (AVARP20010_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP20010_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KDR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KDR. Alternate names: CD309, FLK1, VEGFR, VEGFR2ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-014-62649