Learn More
Sino Biological Recombinant Human GSK-3 beta/GSK3B Protein (Full Length, C199A, His Tag), Active
Catalog No. 502574817
Encompass_Preferred
Recombinant full length human GSK3b (C199A) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag.
Catalog No. 50-257-4817 Supplier Sino Biological Supplier No. G0912H50
Recombinant full length human GSK3b (C199A) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag.