Learn More
Sigma Aldrich Fine Chemicals Biosciences KDS LegatoTM 100 single sy1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7590311EA
Encompass
Infuse only single syringe pump. Accommodates syringes from 0.5 muL to 60 mL. User definable flow rates with selectable target volume or time values to control the total infusion volume.FeaturesHolds one syringe from 0.5 muL to 60 mLStall detectionHigh reSOLUTIONon color touch screenUnparalleled ease of usePassword protected touch displayRun LED light on front panelFull metal chassisBuilt in syringe tableUp to 30 lbs/13 kg linear forceAdvanced microstepping techniquesBuilt in RS485 interfaceUSB portI/O TTL interfaceContinuous mode of operationSpill dam
Catalog No. 50-290-8608