Learn More
Sigma Aldrich Fine Chemicals Biosciences KDS LegatoTM 110 single sy1EA
Catalog No. 502908526
Encompass
Single syringe infuse/ withdraw with program option for 2 programs. 50 steps each. Accomodates syringes 0.5 muL to 60 mL. User definable flow rates with selectable target volume or time values to control the total infusion volume.FeaturesHolds one syringe from 0.5 muL to 60 mLHigh reSOLUTIONon color touch screenUnparalled ease of useTouch pad lock featureLED light on front panelFull metal chassisBuilt in syringe tableUp to 30 lbs linear forceAdvanced microstepping techniquesBuilt in RS485 interfaceUSB portI/O TTL interfaceContinuous mode of operationSpill damProgrammable
Catalog No. 50-290-8526 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7405321EA
Single syringe infuse/ withdraw with program option for 2 programs. 50 steps each. Accomodates syringes 0.5 muL to 60 mL. User definable flow rates with selectable target volume or time values to control the total infusion volume.FeaturesHolds one syringe from 0.5 muL to 60 mLHigh reSOLUTIONon color touch screenUnparalled ease of useTouch pad lock featureLED light on front panelFull metal chassisBuilt in syringe tableUp to 30 lbs linear forceAdvanced microstepping techniquesBuilt in RS485 interfaceUSB portI/O TTL interfaceContinuous mode of operationSpill damProgrammable