Learn More
Sigma Aldrich Fine Chemicals Biosciences KDS LegatoTM 130 single sy1EA
Catalog No. 502908553
Encompass
Single syringe infuse/ withdraw with program option for 2 programs. 50 steps each. Holds one syringes from 0.5 muL to 1000 muL. User definable flow rates with selectable target volume or time values to control the total infusion volume.Features.nbspHolds one syringe from 0.5 muL to 1000 muL.nbspHigh resolution color touch screen.nbspUnparalled ease of use.nbspTouch pad lock feature.nbspLED light on front panel.nbspFull metal chassis.nbspBuilt in syringe table.nbspUp to 11 lbs linear force.nbspAdvanced microstepping techniques.nbspBuilt in RS485 interface.nbspUSB port.nbspI/O TTL interface.nbspContinuous mode of operation.nbspSpill dam
Catalog No. 50-290-8553 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7590821EA
Single syringe infuse/ withdraw with program option for 2 programs. 50 steps each. Holds one syringes from 0.5 muL to 1000 muL. User definable flow rates with selectable target volume or time values to control the total infusion volume.Features.nbspHolds one syringe from 0.5 muL to 1000 muL.nbspHigh resolution color touch screen.nbspUnparalled ease of use.nbspTouch pad lock feature.nbspLED light on front panel.nbspFull metal chassis.nbspBuilt in syringe table.nbspUp to 11 lbs linear force.nbspAdvanced microstepping techniques.nbspBuilt in RS485 interface.nbspUSB port.nbspI/O TTL interface.nbspContinuous mode of operation.nbspSpill dam