Learn More
Sigma Aldrich Fine Chemicals Biosciences KDS LegatoTM 180 dual syri1EA
Catalog No. 502908583
Encompass
Dual syringe infuse/withdraw optimized for low flow with program options for 2 programs. 50 steps each. Accomodates syringes 0.5 muL to 10 mL. User definable flow rates with selectable target volume or time values to control the total infusion volume.FeaturesHolds one or two syringes from 0.5 muL to 10 mLHigh reSOLUTIONon color touch screenUnparalled ease of useTouch pad lock featureLED light on front panelFull metal chassisBuilt in syringe tableUp to 30 lbs linear forceAdvanced microstepping techniquesBuilt in RS485 interfaceUSB portI/O TTL interfaceContinuous mode of operationSpill damProgrammable
Catalog No. 50-290-8583 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7590901EA
Dual syringe infuse/withdraw optimized for low flow with program options for 2 programs. 50 steps each. Accomodates syringes 0.5 muL to 10 mL. User definable flow rates with selectable target volume or time values to control the total infusion volume.FeaturesHolds one or two syringes from 0.5 muL to 10 mLHigh reSOLUTIONon color touch screenUnparalled ease of useTouch pad lock featureLED light on front panelFull metal chassisBuilt in syringe tableUp to 30 lbs linear forceAdvanced microstepping techniquesBuilt in RS485 interfaceUSB portI/O TTL interfaceContinuous mode of operationSpill damProgrammable