Learn More
Sigma Aldrich Fine Chemicals Biosciences KDS LegatoTM 200 series sy1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5695851EA
Encompass
Infuse only dual syringe pump. Accomodates 2 syringes. User definable flow rates with selectable target volume or time values to control the total infusion volume.Features.nbspHolds one or two syringes from 0.5 muL to 140 mL.nbspHigh resolution color touch screen.nbspReal time clock.nbspUnparallel ease of use.nbspTouch pad lock feature.nbspLED light on front panel.nbspFull metal chassis.nbspBuilt in syringe table.nbspUp to 75 lbs linear force.nbspAdvanced microstepping techniques.nbspBuilt in RS485 interface.nbspUSB port.nbspI/O TTL interface.nbspContinuous mode of operation.nbspSpill dam
Catalog No. 50-290-8782