Learn More
Sigma Aldrich Fine Chemicals Biosciences KDS LegatoTM 210P series s1EA
Catalog No. 502908679
Encompass
Programmable Legato 210P dual syringe pump with user defined configurations/programs of up to 800 steps. Up to 40 programs of 20 steps each step can be stored in memory.Features.nbspHolds one or two syringes from 0.5 muL to 140 mL.nbspHigh resolution color touch screen.nbspReal time clock.nbspUnparallel ease of use.nbspTouch pad lock feature.nbspLED light on front panel.nbspFull metal chassis.nbspBuilt in syringe table.nbspUp to 75 lbs linear force.nbspAdvanced microstepping techniques.nbspBuilt in RS485 interface.nbspUSB port.nbspI/O TTL interface.nbspContinuous mode of operation.nbspSpill dam.nbspProgrammable
Catalog No. 50-290-8679 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5696071EA
Add to Cart
Add to Cart
Request a Quote
Programmable Legato 210P dual syringe pump with user defined configurations/programs of up to 800 steps. Up to 40 programs of 20 steps each step can be stored in memory.Features.nbspHolds one or two syringes from 0.5 muL to 140 mL.nbspHigh resolution color touch screen.nbspReal time clock.nbspUnparallel ease of use.nbspTouch pad lock feature.nbspLED light on front panel.nbspFull metal chassis.nbspBuilt in syringe table.nbspUp to 75 lbs linear force.nbspAdvanced microstepping techniques.nbspBuilt in RS485 interface.nbspUSB port.nbspI/O TTL interface.nbspContinuous mode of operation.nbspSpill dam.nbspProgrammable