Learn More
Sigma Aldrich Fine Chemicals Biosciences KDS single syringe pump se1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z4013581EA
Encompass
Series 100 infusion pumps combine precision and simplicity with ease of use and exceptional durablity. The 230 V pumps are CE compliant. Advanced microprocessor-controlled syringe pumps designed especially for low pulse-free flow rates.Easy setup1. Select syringe size2. Enter volume to be dispensed3. Enter flow rate then press start buttonFeaturesHolds any size syringe from 10 muL to 60 mL syringesAutomatic volume control and shutoffLast settings stored in memoryContinuous dispense volume displaySimple menu-driven setup syringe diameter (selected from displayed table) dispense volume and dispense flow rate
Catalog No. 50-290-9127