Learn More
Sigma Aldrich Fine Chemicals Biosciences KDS stainless steel syring1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7598211EA
Encompass
These rugged 316 stainless steel syringes are a perfect complement to the KDS 410 high pressure syringe dispenser. They eliminate the problem of breaking glass syringes when using high pressures. Compatibility with most chemicals is assured as wetted parts are 316 stainless steel Viton(R) for the 20 50 and 100 mL sizes and Perfluoroelastomer for the 8 mL size.Compatible with most syringe pumpsFully autoclavableGood resistance to most chemicalsSafe SOLUTIONon when using high pressure dispensing
Catalog No. 50-290-9105