Learn More
Selleck Chemical LLC KDU691
SDP
Catalog No. 5000002927
Encompass_Preferred
KDU691 is a Plasmodium PI4K-specific inhibitor with IC50 values of 0 18 M and 0 061 M against hypnozoite forms and liver schizontsm respectively
Catalog No. 50-000-02927 Supplier Selleck Chemical LLC Supplier No. S65742MG
KDU691 is a Plasmodium PI4K-specific inhibitor with IC50 values of 0 18 M and 0 061 M against hypnozoite forms and liver schizontsm respectively