Learn More
Medchemexpress LLC HY-12912 5mg Medchemexpress, KDU691 CAS:1513879-19-0 Purity:>98%
SDP
Catalog No. 502081197
Encompass_Preferred
Medchemexpress, HY-12912 5mg KDU691 CAS:1513879-19-0 KDU691, an imidazopyrazine with potent anti-parasitic activity against blood stage schizonts, gametocytes and liver stages, is a Plasmodium PI4K inhibitor. KDU691 selectively inhibits dihydroartemisinin-pretreated Plasmodium falciparum ring-stage parasites. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1197 Supplier Medchemexpress LLC Supplier No. HY129125MG
Medchemexpress, HY-12912 5mg KDU691 CAS:1513879-19-0 KDU691, an imidazopyrazine with potent anti-parasitic activity against blood stage schizonts, gametocytes and liver stages, is a Plasmodium PI4K inhibitor. KDU691 selectively inhibits dihydroartemisinin-pretreated Plasmodium falciparum ring-stage parasites. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.