Learn More
Medchemexpress LLC HY-103583 5mg Medchemexpress, KDU731 CAS:1610610-48-4 Purity:>98%
SDP
Catalog No. 501968129
Encompass_Preferred
Medchemexpress, HY-103583 5mg KDU731 CAS:1610610-48-4 KDU731, an orally active C. parvum PI4K inhibitor with an IC50 value of 25 nM, blocks Cryptosporidium infection in vitro and in vivo. KDU731 is a promising drug candidate for the treatment of diarrhea caused by Cryptosporidium and meets a broad range of safety. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8129 Supplier Medchemexpress LLC Supplier No. HY1035835MG
Medchemexpress, HY-103583 5mg KDU731 CAS:1610610-48-4 KDU731, an orally active C. parvum PI4K inhibitor with an IC50 value of 25 nM, blocks Cryptosporidium infection in vitro and in vivo. KDU731 is a promising drug candidate for the treatment of diarrhea caused by Cryptosporidium and meets a broad range of safety. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.