Learn More
Aviva Systems Biology Corp KEAP1 Antibody - C-terminal region (ARP34728_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34728_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KEAP1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human KEAP1. Alternate names: INrf2, KIAA0132, KLHL19, MGC10630, MGC1114, MGC20887, MGC4407, MGC9454ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-52975