Learn More
Cayman Chemical Keap1humnrecombInt 100ug
SDP
Catalog No. 502950730
Encompass_Preferred
Source: Recombinant human N-terminal His-GST-tagged Keap1 expressed in insect cells • Amino acids: 2-624 (full length) • MW: 97.37 kDa
Catalog No. 50-295-0730 Supplier Cayman Chemical Supplier No. 32035100 UG
Source: Recombinant human N-terminal His-GST-tagged Keap1 expressed in insect cells • Amino acids: 2-624 (full length) • MW: 97.37 kDa