Learn More
Cole-Parmer Keck Ramp Clamp Tubing Clamps, 14 mm, 12/Pk
Supplier:  Cole-Parmer 0683510
Encompass_Preferred
Unique wheel-in-groove clamp design permits one-handed flow control. Simply remove wheel to insert tubing, then roll wheel back into clamp to your desired flow rate. Made of pure PBT, these clamps are safe to 392F (200C). All four sizes accept tubing with a maximum wall thickness of 3/32" (2.4 mm).
Catalog No. NC9992055