Learn More
Chemglass Life Sciences Clamp, Keck, Violet, Spherical Joint, Fits Joint Sizes 12/1, 12/2 & 12/5
SDP
Catalog No. CG145B20
Encompass_Preferred
Clamp, Keck, Violet, Spherical Taper, Fits Joint Sizes 12/1, 12/2 & 12/5. Made of Delrin, these clamps are non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10.
Catalog No. CG145B20 Supplier Chemglass Life Sciences Supplier No. CG145B20
Clamp, Keck, Violet, Spherical Taper, Fits Joint Sizes 12/1, 12/2 & 12/5. Made of Delrin, these clamps are non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10.