Learn More
Chemglass Life Sciences Clamp, Keck, Red, Spherical Joint, Fits Joint Sizes 28/12 & 28/15
SDP
Catalog No. CG145B22
Encompass_Preferred
Clamp, Keck, Red, Standard Taper, Fits Joint Sizes 28/12 & 28/15. Made of Delrin, these clamps are non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10.
Catalog No. CG145B22 Supplier Chemglass Life Sciences Supplier No. CG145B22
Clamp, Keck, Red, Standard Taper, Fits Joint Sizes 28/12 & 28/15. Made of Delrin, these clamps are non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10.