Learn More
Chemglass Life Sciences Clamp, Keck, Dark Yellow, Standard Taper, Fits Joint Sizes 40/35 & 40/50
SDP
Catalog No. CG14508
Encompass_Preferred
Plastic Keck Style Clamp, #24. Stainless Steel clamps to fit 24/40 and 24/25 standard taper joints. Metal clamps are suitable for high temperatures applications.
Catalog No. CG14508 Supplier Chemglass Life Sciences Supplier No. CG14508
Plastic Keck Style Clamp, #24. Stainless Steel clamps to fit 24/40 and 24/25 standard taper joints. Metal clamps are suitable for high temperatures applications.