Learn More
Cole-Parmer Keck Metal Standard Taper Clip Size 24 6/Pk
Supplier:  Cole-Parmer 0683615
Encompass_Preferred
These metal taper clips securely hold conical joints under pressure. Nickel-plated steel taper clips feature superior chemical and temperature resistance—withstand temperatures to 932°>F (500°>C).
Catalog No. NC9401711