Learn More
MSC Kee Double Swivel Socket, 1-1/4 in Pipe, Galvanized, 1 ea
Catalog No. 5000795182
Encompass
This pipe rail double swivel socket is designed for 1-1/4 in pipes, offering a secure and versatile connection for railing systems. Its galvanized finish provides enhanced durability and corrosion resistance for long-lasting performance.
  • Designed for 1-1/4 in pipes
  • Double swivel socket type
  • Constructed from durable iron
  • Features a galvanized finish coating
  • Manufactured in the United Kingdom
Catalog No. 50-007-95182 Supplier MSC Supplier No. 89818629
This pipe rail double swivel socket is designed for 1-1/4 in pipes, offering a secure and versatile connection for railing systems. Its galvanized finish provides enhanced durability and corrosion resistance for long-lasting performance.
  • Designed for 1-1/4 in pipes
  • Double swivel socket type
  • Constructed from durable iron
  • Features a galvanized finish coating
  • Manufactured in the United Kingdom