Learn More
Thermo Scientific Keeper seal wash pump for ICS-3000
Catalog No. 501345183 Shop All Thermo Scientific Products
Encompass
Pump circulates wash solutions through ion chromatography system to remove contaminants, prevent ghost peaks, and clean for maintenance.
Catalog No. 50-134-5183 Supplier Thermo Scientific Supplier No. 062092
Pump circulates wash solutions through ion chromatography system to remove contaminants, prevent ghost peaks, and clean for maintenance.