Learn More
Grainger Keg Dolly 17 1/4 in L 5 1/4 in W
Catalog No. 502751775
Encompass_Preferred
The Grainger 14G297 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for a variety of cleaning and scrubbing tasks. It features an open web construction with non-scratching fibers that are durable and long-lasting. This 8" x 4" hand pad has a medium duty scrubbing surface on both sides for scrubbing pots, pans, dishes, countertops, floors, walls, appliances and more. It effectively removes grease, dirt, food residue and more without damaging surfaces. The pad is latex-free, disposable and ideal for commercial kitchens, restaurants, schools, hospitals and industrial facilities.
Catalog No. 50-275-1775 Supplier Grainger Supplier No. 14G297
The Grainger 14G297 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for a variety of cleaning and scrubbing tasks. It features an open web construction with non-scratching fibers that are durable and long-lasting. This 8" x 4" hand pad has a medium duty scrubbing surface on both sides for scrubbing pots, pans, dishes, countertops, floors, walls, appliances and more. It effectively removes grease, dirt, food residue and more without damaging surfaces. The pad is latex-free, disposable and ideal for commercial kitchens, restaurants, schools, hospitals and industrial facilities.