Learn More
Grainger Vestil Keg Lifter Kit, Steel, 400 lb. Capacity, 1 ea
Catalog No. 502774806
Encompass_Preferred
This keg lifter kit serves as an add-on attachment designed to facilitate the efficient transport and maneuvering of kegs. The unit is constructed from steel and is intended for use with the PEL-400S model.
  • Keg lift attachment for simple transferring
  • Strong construction for long life
  • Compact design for storage
  • Easy to wash down
Catalog No. 50-277-4806 Supplier Grainger Supplier No. 172X91
This keg lifter kit serves as an add-on attachment designed to facilitate the efficient transport and maneuvering of kegs. The unit is constructed from steel and is intended for use with the PEL-400S model.
  • Keg lift attachment for simple transferring
  • Strong construction for long life
  • Compact design for storage
  • Easy to wash down