Learn More
Analytical Sales & Services Kel-F Adapter, 1/4-28 Male to 1/2-20 Female
Catalog No. 5000804524
Encompass
Adapter, 1/2-20 F (for 1/4" OD Tubing) to 1/4-28 M\n\nPolypropylene\n\nQty/Pkg: Each
Catalog No. 50-008-04524 Supplier Analytical Sales & Services Supplier No. 59684
Adapter, 1/2-20 F (for 1/4" OD Tubing) to 1/4-28 M\n\nPolypropylene\n\nQty/Pkg: Each