Learn More
Ace Glass, Inc. KEL-F-HUB ADA 40 IN. LGTH Kel-F-Hub Ada 40 In. Lgth
SDP
Catalog No. 50972365
Encompass
Needle adapter designed to eliminate dead space between end of standard luer taper and bottom of standard hub. Needle extends entirely through hub and fits into counter-bored hole in the Luer male, and is seated by the hub.
Catalog No. 50-972-365 Supplier Ace Glass, Inc. Supplier No. 585610
Needle adapter designed to eliminate dead space between end of standard luer taper and bottom of standard hub. Needle extends entirely through hub and fits into counter-bored hole in the Luer male, and is seated by the hub.