Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kelch-like ECH-associated protein 1 | >90.0% | 100 UG
SDP

Catalog No. 5000290775
Encompass_Preferred

Recombinant human Kelch-like ECH-associated protein 1 (KEAP1) produced in Sf9 insect cells and supplied as a 0.22 μm filtered solution in 20 mM Tris, 300 mM NaCl, 10% glycerol, pH 7.5. The protein contains N-terminal His and GST tags, is supplied at laboratory scale (100 μg), and is intended for biochemical and binding assays. Store frozen and avoid repeated freeze-thaw cycles; buffer exchange is recommended for SPR applications.

  • Recombinant human protein expressed in Sf9 insect cells.
  • N-terminal His and GST tags facilitate purification and detection.
  • Supplied as a filtered solution in Tris/NaCl/glycerol buffer.
  • Observed molecular weight ~93-109 kDa by SDS-PAGE.
  • Stable when stored at -80 °C; avoid repeated freeze-thaw cycles.

Catalog No. 50-002-90775 Supplier Medchemexpress LLC Supplier No. HYP75899100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Kelch-like ECH-associated protein 1 (KEAP1) produced in Sf9 insect cells and supplied as a 0.22 μm filtered solution in 20 mM Tris, 300 mM NaCl, 10% glycerol, pH 7.5. The protein contains N-terminal His and GST tags, is supplied at laboratory scale (100 μg), and is intended for biochemical and binding assays. Store frozen and avoid repeated freeze-thaw cycles; buffer exchange is recommended for SPR applications.

  • Recombinant human protein expressed in Sf9 insect cells.
  • N-terminal His and GST tags facilitate purification and detection.
  • Supplied as a filtered solution in Tris/NaCl/glycerol buffer.
  • Observed molecular weight ~93-109 kDa by SDS-PAGE.
  • Stable when stored at -80 °C; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.