Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kelch-like ECH-associated protein 1 (KEAP1) | ≥90.0% | 50 UG
SDP

Catalog No. 5000292209
Encompass_Preferred

Recombinant human Kelch-like ECH-associated protein 1 (KEAP1) produced in Sf9 insect cells with N-terminal His and GST affinity tags. Supplied as a 0.22 μm-filtered solution in 20 mM Tris, 300 mM NaCl, 10% glycerol (pH 7.5), this protein is designed for biochemical and binding assays involving the NRF2-KEAP1 pathway.

  • Recombinant full-length human protein expressed in Sf9 insect cells.
  • N-terminal His and GST affinity tags for purification and detection.
  • Supplied in buffered, filtered solution (20 mM Tris, 300 mM NaCl, 10% glycerol, pH 7.5).
  • Purity ≥90.0% as determined by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg by LAL method.
  • Apparent molecular weight ~93-109 kDa by SDS-PAGE.
  • Store at -80°C for long-term stability; avoid repeated freeze-thaw cycles.

Catalog No. 50-002-92209 Supplier Medchemexpress LLC Supplier No. HYP7589950UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Kelch-like ECH-associated protein 1 (KEAP1) produced in Sf9 insect cells with N-terminal His and GST affinity tags. Supplied as a 0.22 μm-filtered solution in 20 mM Tris, 300 mM NaCl, 10% glycerol (pH 7.5), this protein is designed for biochemical and binding assays involving the NRF2-KEAP1 pathway.

  • Recombinant full-length human protein expressed in Sf9 insect cells.
  • N-terminal His and GST affinity tags for purification and detection.
  • Supplied in buffered, filtered solution (20 mM Tris, 300 mM NaCl, 10% glycerol, pH 7.5).
  • Purity ≥90.0% as determined by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg by LAL method.
  • Apparent molecular weight ~93-109 kDa by SDS-PAGE.
  • Store at -80°C for long-term stability; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.