Learn More
Electron Microscopy Sciences EMS Kelley Forceps Premium Grade, Curved
SDP
Catalog No. 501931461
Encompass_Preferred
These stainless steel forceps feature a finger ring grip and adjustable interlocking cross arms for tight clamping of tissue.
Tips: Curved
OAL: 5-1/2"
This item is not returnable. View return policy
Catalog No. 50-193-1461 Supplier Electron Microscopy Sciences Supplier No. 7299930
These stainless steel forceps feature a finger ring grip and adjustable interlocking cross arms for tight clamping of tissue.
Tips: Curved
OAL: 5-1/2"