Learn More
Electron Microscopy Sciences Kellner Eyepiece, X10
SDP
Catalog No. 5025833
Encompass_Preferred
Focusing eyepiece fits most standard monocular microscope with eyepiece graticules of 16mm diameter. Composed of a convex lens and an achromatic lens so the graticule is brought into sharp focus when superimposed on the object image.
Catalog No. 50-258-33 Supplier Electron Microscopy Sciences Supplier No. 67990F10
Focusing eyepiece fits most standard monocular microscope with eyepiece graticules of 16mm diameter. Composed of a convex lens and an achromatic lens so the graticule is brought into sharp focus when superimposed on the object image.