Learn More
Electron Microscopy Sciences Kellner Wide-Field Eyepiece, X10
SDP
Catalog No. 5025832
Encompass_Preferred
Focusing eyepiece with high eye point (useful for spectacle wearers). Non -focusing wide field.
Catalog No. 50-258-32 Supplier Electron Microscopy Sciences Supplier No. 67990F12
Focusing eyepiece with high eye point (useful for spectacle wearers). Non -focusing wide field.