Learn More
World Precision Instrument Kelly Forceps, 14cm, Straight, 12-pack
SDP
Catalog No. 502539456
Encompass
Kelly hemostatic forceps are used to clamp blood vessels and hold heavy tissues in place. Kelly forceps, both straight and curved, are available. Our forceps feature a locking ratchet and wider finger rings for easy handling.
Catalog No. 50-253-9456 Supplier World Precision Instrument Supplier No. 50370212
Kelly hemostatic forceps are used to clamp blood vessels and hold heavy tissues in place. Kelly forceps, both straight and curved, are available. Our forceps feature a locking ratchet and wider finger rings for easy handling.