Learn More
Stoelting™ Kelly Forceps
SDP
Catalog No. 10001332 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Curved
Curved, Left-handed
Straight
Straight, Left-handed
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-001-332 Straight
10-001-333 Straight, Left-handed
10-001-334 Curved
10-001-335 Curved, Left-handed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-001-332 Supplier Stoelting™ Supplier No. 5211014
Only null left
Add to Cart
Add to Cart
Kelly Forceps are used in many surgical procedures usually to control bleeding or the flow of liquids in tubing.
  • Scissor-like ring handles just like scissors
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
Specifications
For Use With (Application) Neuroscience
Length (English) 5.5 in.
Model Straight