Learn More
Stoelting™ Kelly Forceps
SDP
Catalog No. 10001334 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Curved
Curved, Left-handed
Straight
Straight, Left-handed
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-001-334 Curved
10-001-332 Straight
10-001-333 Straight, Left-handed
10-001-335 Curved, Left-handed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-001-334 Supplier Stoelting™ Supplier No. 5211016
Add to Cart
Add to Cart
Kelly Forceps are used in many surgical procedures usually to control bleeding or the flow of liquids in tubing.
  • Scissor-like ring handles just like scissors
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
Specifications
For Use With (Application) Neuroscience
Length (English) 5.5 in.
Model Curved