Learn More
Med Vet International Miltex Vantage Kelly Hemostatic Forceps, 5.5in, Straight
Catalog No. 502522264
Encompass_Preferred
The Miltex Vantage V97-36 Kelly forceps are made from high quality Japanese stainless steel and measure 5". They have a finger ring handle with a ratchet lock and straight tips with serrations.
Catalog No. 50-252-2264 Supplier Med Vet International Supplier No. MILV9736
The Miltex Vantage V97-36 Kelly forceps are made from high quality Japanese stainless steel and measure 5". They have a finger ring handle with a ratchet lock and straight tips with serrations.