Learn More
Sklar Instruments Sklarlite Kelly Hemostatic Forceps, Sklar, 5-1/2 in., Blunt, Straight, Serrated, Extra Light, Stainless Steel. EA/1.
SDP
Catalog No. 501437955 Shop All Sklar Instruments Products
Encompass_Preferred
Sklarlite Kelly Hemostatic Forceps, Sklar, 5-1/2 in., Blunt, Straight, Serrated, Extra Light, Stainless Steel. Sklarlite Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. Sklarlite instruments are a lighter version of standard Sklar instruments. This lightweight modification provides the surgeon with greater control and less fatigue. This product is straight with serrated tips and a length of 5-1/2 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7955 Supplier Sklar Instruments Supplier No. 232107
Sklarlite Kelly Hemostatic Forceps, Sklar, 5-1/2 in., Blunt, Straight, Serrated, Extra Light, Stainless Steel. Sklarlite Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. Sklarlite instruments are a lighter version of standard Sklar instruments. This lightweight modification provides the surgeon with greater control and less fatigue. This product is straight with serrated tips and a length of 5-1/2 inches.