Learn More
Sklar Instruments Kelly Hemostatic Forceps, 5-1/2 in., Curved, Serrated, Stainless Steel, EA.
SDP
Catalog No. 501109743 Shop All Sklar Instruments Products
Encompass_Preferred
Kelly Hemostatic Forceps, 5-1/2 in., Curved, Serrated, Stainless Steel, EA. Sklar Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. This product is curved with serrated tips and a length of 5-1/2 inches.
Catalog No. 50-110-9743 Supplier Sklar Instruments Supplier No. 172155
Kelly Hemostatic Forceps, 5-1/2 in., Curved, Serrated, Stainless Steel, EA. Sklar Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. This product is curved with serrated tips and a length of 5-1/2 inches.