Learn More
Sklar Instruments Forceps, Surgi-OR Kelly-Rankin Forceps, Sklar 6-1/4 in., Straight, Serrated, Stainless Steel, EA.
SDP
Catalog No. 501126514 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Surgi-OR Kelly-Rankin Forceps, Sklar, 6-1/4 in., Straight, Serrated, Stainless Steel, EA. Surgi-OR Kelly-Rankin Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. The jaws are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. Standard Kelly forceps are available in two lengths, 5-1/2 inches and 7 inches. Kelly forceps that have an overall length of 6-1/4 inches are referred to as Kelly-Rankin forceps. Surgi-OR products are mid-grade instruments suitable for the operating room and other surgical centers. This product is straight with serrated jaws and a length of 6-1/4 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-112-6514 Supplier Sklar Instruments Supplier No. 95465
Forceps, Surgi-OR Kelly-Rankin Forceps, Sklar, 6-1/4 in., Straight, Serrated, Stainless Steel, EA. Surgi-OR Kelly-Rankin Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. The jaws are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. Standard Kelly forceps are available in two lengths, 5-1/2 inches and 7 inches. Kelly forceps that have an overall length of 6-1/4 inches are referred to as Kelly-Rankin forceps. Surgi-OR products are mid-grade instruments suitable for the operating room and other surgical centers. This product is straight with serrated jaws and a length of 6-1/4 inches. Lifetime warranty against all manufacturer defects.