Learn More
Cayman Chemical KEMPFPKYPVEP Peptide (bovine, ovine) (trifluoroacetate salt), Purity: ≥95%, 250 mg
SDP
Catalog No. 502918539
Encompass_Preferred
A peptide fragment of β-casein; can be produced by the protease M from A. oryzae and is found in dairy products fermented by fungi but not bacteria; increases the levels of dopamine, norepinephrine, and homovanillic acid, but not serotonin, in the frontal cortex of mice at 2 mg/kg; improves spatial alternation retention in the Y-maze spatial alternation task and increases novel object exploration time in a novel object recognition test in a mouse model of scopolamine-induced amnesia at 2 mg/kg
Catalog No. 50-291-8539 Supplier Cayman Chemical Supplier No. 38850250 MG
A peptide fragment of β-casein; can be produced by the protease M from A. oryzae and is found in dairy products fermented by fungi but not bacteria; increases the levels of dopamine, norepinephrine, and homovanillic acid, but not serotonin, in the frontal cortex of mice at 2 mg/kg; improves spatial alternation retention in the Y-maze spatial alternation task and increases novel object exploration time in a novel object recognition test in a mouse model of scopolamine-induced amnesia at 2 mg/kg