Learn More
CPC Scientific H-Leu-Arg-Arg-Ala-Ser-Leu-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP015A
Encompass
Kemptide is a synthetic peptide substrate corresponding to part of the phosphyorylation site sequence in porcine liver pyruvate kinase. It is phosphorylated in vitro and in vivo by cAMP-dependent protein kinase and in vitro by protein kinase C.SEQUENCE: H-Leu-Arg-Arg-Ala-Ser-Leu-Gly-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: LRRASLGMOLECULAR FORMULA: C32H61N13O9MOLECULAR WEIGHT: 771.92STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [65189-71-1]SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:B.E. Kemp et al., Fed. Proc., 35, 1384 (1976)SKU(s): PKCP-015A
Catalog No. 50-211-0167