Learn More
Apexbio Technology LLC KEMPTIDE-ACETATE-SALT-10MG
SDP
Catalog No. 5000567463
Encompass_Preferred
Kemptide acetate salt is a synthetic peptide used extensively in biomedical research particularly in studies involving kinase activity Originating as a substrate for protein kinase A (PKA) Kemptide s mechanism of action involves the phosphorylation of threonine residues by serine/threonine-specific kinases This phosphorylation event is crucial for studying signal transduction pathways and enzyme-substrate interactions It exhibits in vitro activity primarily in the nanomolar to low micromolar range Kemptide is commonly used in various experimental setups including cell-based assays and in vitro kinase assays to investigate enzyme kinetics and for drug screening purposes Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67463 Supplier Apexbio Technology LLC Supplier No. C858310
Kemptide acetate salt is a synthetic peptide used extensively in biomedical research particularly in studies involving kinase activity Originating as a substrate for protein kinase A (PKA) Kemptide s mechanism of action involves the phosphorylation of threonine residues by serine/threonine-specific kinases This phosphorylation event is crucial for studying signal transduction pathways and enzyme-substrate interactions It exhibits in vitro activity primarily in the nanomolar to low micromolar range Kemptide is commonly used in various experimental setups including cell-based assays and in vitro kinase assays to investigate enzyme kinetics and for drug screening purposes Experimental concentrations typically depend on the specific research design