Learn More
Bachem Kemptide Acetate
SDP
Supplier:  Bachem 4095917.0005
Encompass_Preferred
Kemptide Acetate, H-Leu-Arg-Arg-Ala-Ser-Leu-Gly-OH, 65189-71-1, C32H61N13O9, Mr 771.92, 5mg. Kemptide is a synthetic peptide substrate corresponding to part of the phosphyorylation site sequence in porcine liver pyruvate kinase.
Catalog No. 50-194-7494