Learn More
CPC Scientific 5TAMRA-Leu-Arg-Arg-Ala-Ser-Leu-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PROK001B
Encompass
A phosphate acceptor peptide that serves as a synthetic substrate for PKA (Km = 16 ?M). The corresponding fluorescent and biotinylated peptides are also proven to be good substrates for PKA.SEQUENCE: 5TAMRA-Leu-Arg-Arg-Ala-Ser-Leu-Gly-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: 5TAMRA-LRRASLGMOLECULAR FORMULA: C57H81N15O13MOLECULAR WEIGHT: 1184.4STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:Pfister, TD. and KB. Storey, Insect Biochem. Mol. Biol. 32, 505 (2002)Zelada, A. et al. Eur. J. Biochem. 252, 245 (1998)SKU(s): PROK-001A, PROK-001B
Catalog No. 50-211-0222