Learn More
Bachem Kemptide Trifluoroacetate
SDP
Supplier:  Bachem 4010606.0025
Encompass_Preferred
Kemptide Trifluoroacetate, H-Leu-Arg-Arg-Ala-Ser-Leu-Gly-OH, 65189-71-1, C32H61N13O9, Mr 771.92, 25mg. Kemptide, LRRASLG, is a synthetic peptide substrate corresponding to part of the phosphorylation site sequence in porcine liver pyruvate kinase. Kemptide is phosphorylated in vitro and in vivo by cAMP-dependent protein kinase and in vitro by protein kinase C.
Catalog No. 50-194-7495